5-Phenylpyrimidin-2-amine 97%

Supplier: Ambeed
Warning

A474301-1G A474301-250MG
77143-712EA 140.07 USD
77143-712 77143-714
5-Phenylpyrimidin-2-amine 97%
5-Phenylpyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉N₃
MW: 171.19 g/mol
MDL Number: MFCD01646362
CAS Number: 31408-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR