tert-Butyl 5-methoxyisoindoline-2-carboxylate 97%

Supplier: Ambeed
Warning

A473514-1G A473514-250MG
77143-706EA 268.41 USD
77143-706 77143-708
tert-Butyl 5-methoxyisoindoline-2-carboxylate 97%
tert-Butyl 5-methoxyisoindoline-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₃
MW: 249.30 g/mol
MDL Number: MFCD16620885
CAS Number: 1824073-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR