1-Hexyl-2,3-dimethylimidazolium bromide 95+%

Supplier: Ambeed
Warning

Synonyms: 1-Hexyl-2,3-dimethyl-1H-imidazol-3-ium bromide

A472891-100G A472891-25G A472891-5G
77143-700EA 172.93 USD
77143-700 77143-702 77143-704
1-Hexyl-2,3-dimethylimidazolium bromide 95+%
1-Hexyl-2,3-dimethylimidazolium bromide

Precaución: for Research Use Only

Formula: C₁₁H₂₁BrN₂
MW: 261.21 g/mol
MDL Number: MFCD03427616
CAS Number: 411222-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR