4-(1,2,3,4-Tetrahydro-2-isoquinolinylmethyl)benzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)-1,2,3,4-tetrahydroisoquinoline, 2-[4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzyl]-1,2,3,4-tetrahydro-isoquinoline

A470411-100MG A470411-1G A470411-250MG A470411-5G
77143-670EA 25.76 USD
77143-670 77143-672 77143-674 77143-676
4-(1,2,3,4-Tetrahydro-2-isoquinolinylmethyl)benzeneboronic acid pinacol ester 98%
4-(1,2,3,4-Tetrahydro-2-isoquinolinylmethyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₂H₂₈BNO₂
MDL Number: MFCD16294539
CAS Number: 1315281-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR