(S)-Tetrahydrofuran-3-yl 3-(3-(3-methoxy-4-(oxazol-5-yl)phenyl)ureido)benzylcarbamate 98%

Supplier: Ambeed
Warning

Synonyms: Merimepodib

A470058-10MG A470058-25MG A470058-2MG A470058-50MG A470058-5MG A470058-1MG
77143-658EA 333.13 USD
77143-658 77143-660 77143-662 77143-664 77143-666 77686-251
(S)-Tetrahydrofuran-3-yl 3-(3-(3-methoxy-4-(oxazol-5-yl)phenyl)ureido)benzylcarbamate 98%
(S)-Tetrahydrofuran-3-yl 3-(3-(3-methoxy-4-(oxazol-5-yl)phenyl)ureido)benzylcarbamate

Precaución: for Research Use Only

Formula: C₂₃H₂₄N₄O₆
MW: 452.45 g/mol
MDL Number: MFCD09837807
CAS Number: 198821-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR