2-((1H-Indol-3-yl)methylene)malononitrile 98%

Supplier: Ambeed
Warning

A469194-100MG A469194-1G A469194-250MG
77143-642EA 18.54 USD
77143-642 77143-644 77143-646
2-((1H-Indol-3-yl)methylene)malononitrile 98%
2-((1H-Indol-3-yl)methylene)malononitrile

Precaución: for Research Use Only

Formula: C₁₂H₇N₃
MW: 193.20 g/mol
MDL Number: MFCD00022740
CAS Number: 75629-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR