(2-Methoxy-4,6-bis(trifluoromethyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-4,6-bis(trifluoromethyl)phenylboronic acid

A467607-100MG A467607-250MG
77143-636EA 64.72 USD
77143-636 77143-638
(2-Methoxy-4,6-bis(trifluoromethyl)phenyl)boronic acid 97%
(2-Methoxy-4,6-bis(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₇BF₆O₃
MW: 287.95 g/mol
MDL Number: MFCD27756648
CAS Number: 1067228-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR