tert-Butyl 4-(4-chloropyridin-2-yl)piperazine-1-carboxylate 97%
Supplier: Ambeed
|
Danger
|
A467437-100MG
A467437-1G
A467437-250MG
77143-630EA
68.96
USD
77143-630
77143-632
77143-634
tert-Butyl 4-(4-chloropyridin-2-yl)piperazine-1-carboxylate 97%
tert-Butyl 4-(4-chloropyridin-2-yl)piperazine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₄H₂₀ClN₃O₂ MW: 297.78 g/mol |
MDL Number:
MFCD14702604 CAS Number: 936368-68-2 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...