(S,S)-(-)-Bis(ɑ-methylbenzyl)amine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (S,S)-(-)-Bis(1-phenylethyl)amine Hydrochloride, (S)-Bis((S)-1-phenylethyl)amine hydrochloride

A466721-1G A466721-250MG A466721-25G A466721-5G
77143-622EA 25.75 USD
77143-622 77143-624 77143-626 77143-628
(S,S)-(-)-Bis(ɑ-methylbenzyl)amine hydrochloride 98%
(S,S)-(-)-Bis(alpha-methylbenzyl)amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₁₉N
MDL Number: MFCD00216671
CAS Number: 40648-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR