(R)-(-)-4-Phenyl-2-butanol 98%

Supplier: Ambeed
Danger

Synonyms: (R)-(-)-4-Phenylbutan-2-ol, (R)-4-Phenylbutan-2-ol

A464763-5G A464763-1G
77143-614EA 183.54 USD
77143-614 77594-464
(R)-(-)-4-Phenyl-2-butanol 98%
(R)-(-)-4-Phenyl-2-butanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
MDL Number: MFCD03093081
CAS Number: 39516-03-5
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR