5-Amino-2-(trifluoromethyl)benzonitrile 97%
Supplier: Ambeed
|
Warning
|
A463863-250MG
A463863-5G
77143-610EA
28.84
USD
77143-610
77143-612
5-Amino-2-(trifluoromethyl)benzonitrile 97%
5-Amino-2-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only
Formula:
C₈H₅F₃N₂ |
MDL Number:
MFCD20694851 CAS Number: 354814-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...