(S)-2,3-Bis(benzyloxy)propan-1-ol 95%

Supplier: Ambeed
Warning

A463322-1G A463322-5G
77143-604EA 166.57 USD
77143-604 77143-606
(S)-2,3-Bis(benzyloxy)propan-1-ol 95%
(S)-2,3-Bis(benzyloxy)propan-1-ol

Precaución: for Research Use Only

Formula: C₁₇H₂₀O₃
MW: 272.33 g/mol
MDL Number: MFCD02683556
CAS Number: 20196-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR