(6-(4-Fluorophenoxy)pyridin-3-yl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A462859-1G
A462859-250MG
A462859-5G
77143-598EA
156.57
USD
77143-598
77143-600
77143-602
(6-(4-Fluorophenoxy)pyridin-3-yl)boronic acid 98%
(6-(4-Fluorophenoxy)pyridin-3-yl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₁H₉BFNO₃ |
MDL Number:
MFCD22375013 CAS Number: 1973462-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...