1-(5-Phenylpyridin-3-yl)piperazine 97%

Supplier: Ambeed
Warning

A461758-100MG A461758-250MG
77143-592EA 46.35 USD
77143-592 77143-594
1-(5-Phenylpyridin-3-yl)piperazine 97%
1-(5-Phenylpyridin-3-yl)piperazine

Precaución: for Research Use Only

Formula: C₁₅H₁₇N₃
MDL Number: MFCD26390882
CAS Number: 223794-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR