[5-Phenyl-3-isoxazolyl]methanol 98%

Supplier: Ambeed
Warning

Synonyms: (5-Phenylisoxazol-3-yl)methanol, 3-Hydroxymethyl-5-phenylisoxazole

A459729-100MG A459729-1G A459729-250MG A459729-5G
77143-572EA 42.44 USD
77143-572 77143-574 77143-576 77143-578
[5-Phenyl-3-isoxazolyl]methanol 98%
[5-Phenyl-3-isoxazolyl]methanol

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.1840 g/mol
MDL Number: MFCD01444104
CAS Number: 1619-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR