tert-Butyl (7-bromoheptyl)carbamate 95%

Supplier: Ambeed
Warning

A459503-1G A459503-250MG A459503-100MG
77143-570EA 88.58 USD
77143-570 77551-398 77616-988
tert-Butyl (7-bromoheptyl)carbamate 95%
tert-Butyl (7-bromoheptyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₂₄BrNO₂
MDL Number: MFCD22576425
CAS Number: 142356-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR