2-Bromo-5-(trifluoromethyl)phenylhydrazine hydrochloride 98%

Supplier: Ambeed
Warning

A459008-10G A459008-1G A459008-250MG A459008-5G
77143-560EA 271.6 USD
77143-560 77143-562 77143-564 77143-566
2-Bromo-5-(trifluoromethyl)phenylhydrazine hydrochloride 98%
2-Bromo-5-(trifluoromethyl)phenylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₆BrF₃N₂·ClH
MW: 291.496 g/mol
MDL Number: MFCD03094412
CAS Number: 529512-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR