(S)-1-((1-Methylpyrrolidin-2-yl)methyl)piperidine 98%

Supplier: Ambeed
Warning

A457206-1G A457206-5G A457206-250MG
77143-550EA 73.21 USD
77143-550 77143-552 77746-933
(S)-1-((1-Methylpyrrolidin-2-yl)methyl)piperidine 98%
(S)-1-((1-Methylpyrrolidin-2-yl)methyl)piperidine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂
MW: 182.31 g/mol
MDL Number: MFCD18642280
CAS Number: 84466-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR