3-Bromo-4-nitrobenzotrifluoride 97%

Supplier: Ambeed
Warning

A456578-1G A456578-250MG
77143-544EA 24.72 USD
77143-544 77143-546
3-Bromo-4-nitrobenzotrifluoride 97%
3-Bromo-4-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃BrF₃NO₂
MDL Number: MFCD09998160
CAS Number: 132839-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR