5-Bromo-2-hydroxybenzeneboronic acid 98%

Supplier: Ambeed
Warning

A455096-250MG A455096-5G
77143-542EA 25.75 USD
77143-542 77746-928
5-Bromo-2-hydroxybenzeneboronic acid 98%
5-Bromo-2-hydroxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BBrO₃
MDL Number: MFCD00093408
CAS Number: 89598-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR