2-Bromo-4,5-dimethylpyridine 97%

Supplier: Ambeed
Warning

A450895-100MG A450895-1G A450895-5G
77143-530EA 29.87 USD
77143-530 77143-532 77143-534
2-Bromo-4,5-dimethylpyridine 97%
2-Bromo-4,5-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MDL Number: MFCD11869962
CAS Number: 66533-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR