5-Bromo-2-methylbenzotrifluoride 98%

Supplier: Ambeed
Warning

A450827-1G A450827-25G A450827-5G
77143-524EA 6.37 USD
77143-524 77143-526 77143-528
5-Bromo-2-methylbenzotrifluoride 98%
5-Bromo-2-methylbenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆BrF₃
MW: 239.04 g/mol
MDL Number: MFCD01631587
CAS Number: 86845-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR