(2-Butyl-5-chloro-1H-imidazol-4-yl)methanol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Butyl-5-chloro-4-hydroxymethylimidazole, (2-Butyl-4-chloro-1H-imidazol-5-yl)methanol

A449084-1G A449084-250MG A449084-25G A449084-5G
77143-506EA 43.5 USD
77143-506 77143-508 77143-510 77143-512
(2-Butyl-5-chloro-1H-imidazol-4-yl)methanol 98%
(2-Butyl-5-chloro-1H-imidazol-4-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₁₃ClN₂O
MW: 188.66 g/mol
MDL Number: MFCD01934397
CAS Number: 79047-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR