(-)-Dipivaloyl-L-tartaric acid 96%

Supplier: Ambeed
Warning

Synonyms: (2R,3R)-2,3-Bis(pivaloyloxy)succinic acid, (-)-O,O'-Dipivaloyl-L-tartaric acid, (-)-Bis(2,2-dimethylpropionyl)-L-tartaric Acid

A447985-100G A447985-10G A447985-250MG A447985-25G A447985-5G
77143-496EA 127.72 USD
77143-496 77143-498 77143-500 77143-502 77143-504
(-)-Dipivaloyl-L-tartaric acid 96%
(-)-Dipivaloyl-L-tartaric acid

Precaución: for Research Use Only

Formula: C₁₄H₂₂O₈
MDL Number: MFCD00066535
CAS Number: 65259-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR