(S,S)-(+)-2,5-Hexanediol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S,5S)-2,5-Hexanediol, (2S,5S)-Hexane-2,5-diol, (2S,5S)-(+)-2,5-Hexanediol, (S,S)-(+)-2,5-Dihydroxy hexane
A447376-1G
A447376-250MG
A447376-5G
77143-490EA
25.47
USD
77143-490
77143-492
77143-494
(S,S)-(+)-2,5-Hexanediol 98%
(S,S)-(+)-2,5-Hexanediol
Precaución: for Research Use Only
Formula:
C₆H₁₄O₂ MW: 118.18 g/mol |
MDL Number:
MFCD00082583 CAS Number: 34338-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...