2-Chloro-4-fluorobenzylamine 97%

Supplier: Ambeed
Danger

A446544-10G A446544-1G A446544-25G A446544-5G
77143-476EA 24.41 USD
77143-476 77143-478 77143-480 77143-482
2-Chloro-4-fluorobenzylamine 97%
2-Chloro-4-fluorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₇ClFN
MW: 159.59 g/mol
MDL Number: MFCD00042532
CAS Number: 15205-11-5
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR