2-Fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzamide 95%

Supplier: Ambeed
Warning

A444758-100MG A444758-250MG
77143-442EA 73.21 USD
77143-442 77143-444
2-Fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzamide 95%
2-Fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzamide

Precaución: for Research Use Only

Formula: C₁₆H₁₂FN₃O₂
MW: 297.28 g/mol
MDL Number: MFCD29918469
CAS Number: 1956340-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR