1-(2-Fluorophenyl)-2-thiourea 95%

Supplier: Ambeed
Danger

A444470-1G A444470-25G
77143-428EA 10.3 USD
77143-428 77143-430
1-(2-Fluorophenyl)-2-thiourea 95%
1-(2-Fluorophenyl)-2-thiourea

Precaución: for Research Use Only

Formula: (C₆H₄F)NC(S)NH₂
MDL Number: MFCD00041148
CAS Number: 656-32-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR