4-Bromo-3-(hydroxymethyl)-1-methyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: (4-Bromo-1-methyl-1H-pyrazol-3-yl)methanol

A443991-1G A443991-250MG A443991-5G
77143-422EA 51.99 USD
77143-422 77143-424 77143-426
4-Bromo-3-(hydroxymethyl)-1-methyl-1H-pyrazole 98%
4-Bromo-3-(hydroxymethyl)-1-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₇BrN₂O
MW: 191.026 g/mol
MDL Number: MFCD06202888
CAS Number: 915707-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR