trans-3-Bromocinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: (E)-3-(3-Bromophenyl)acrylic acid

A443472-100G A443472-1G A443472-25G A443472-500G A443472-5G
77143-396EA 192.03 USD
77143-396 77143-398 77143-400 77143-402 77143-404
trans-3-Bromocinnamic acid 98%
trans-3-Bromocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₇BrO₂
MW: 227.06 g/mol
MDL Number: MFCD00004382
CAS Number: 14473-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR