2-((5-Bromo-4-(4-cyclopropylnaphthalen-1-yl)-4H-1,2,4-triazol-3-yl)thio)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: Lesinurad

A440574-10MG A440574-1MG A440574-50MG A440574-5MG A440574-250MG A440574-5G
77143-382EA 8.49 USD
77143-382 77143-384 77143-386 77143-388 77572-376 77686-195
2-((5-Bromo-4-(4-cyclopropylnaphthalen-1-yl)-4H-1,2,4-triazol-3-yl)thio)acetic acid 98%
2-((5-Bromo-4-(4-cyclopropylnaphthalen-1-yl)-4H-1,2,4-triazol-3-yl)thio)acetic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₄BrN₃O₂S
MW: 404.28 g/mol
MDL Number: MFCD22572730
CAS Number: 878672-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR