2-(2-Cyanophenoxy)acetic acid 97%

Supplier: Ambeed
Warning

A437967-10G A437967-1G A437967-5G
77143-344EA 83.43 USD
77143-344 77143-346 77143-348
2-(2-Cyanophenoxy)acetic acid 97%
2-(2-Cyanophenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MDL Number: MFCD02257373
CAS Number: 6574-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR