Indium(I) chloride 97%

Supplier: Ambeed
Warning

A437072-10G A437072-1G A437072-5G
77143-338EA 114.33 USD
77143-338 77143-340 77143-342
Indium(I) chloride 97%
Indium(I) chloride

Precaución: for Research Use Only

Formula: InCl
MDL Number: MFCD00148875
CAS Number: 13465-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR