N-Carbobenzyloxy-D-serine-beta-lactone 95%

Supplier: Ambeed
Warning

A436970-100MG A436970-1G A436970-250MG A436970-5G
77143-330EA 15.92 USD
77143-330 77143-332 77143-334 77143-336
N-Carbobenzyloxy-D-serine-beta-lactone 95%
N-Carbobenzyloxy-D-serine-beta-lactone

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₄
MW: 221.21 g/mol
MDL Number: MFCD11519130
CAS Number: 98632-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR