tert-Butyl (2-chloropyrimidin-4-yl)carbamate 95%
Supplier: Ambeed
|
Warning
|
A436555-1G
A436555-250MG
A436555-5G
77143-324EA
48.41
USD
77143-324
77143-326
77143-328
tert-Butyl (2-chloropyrimidin-4-yl)carbamate 95%
tert-Butyl (2-chloropyrimidin-4-yl)carbamate
Precaución: for Research Use Only
Formula:
C₉H₁₂ClN₃O₂ |
MDL Number:
MFCD16036511 CAS Number: 849751-48-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...