(S)-(-)-N,N-Dimethyl-1-phenylethylamine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-alpha-Methyl-N,N-dimethylbenzylamine, (S)-N,N-Dimethyl-1-phenylethanamine

A431408-1G A431408-5G
77143-280EA 60.48 USD
77143-280 77616-862
(S)-(-)-N,N-Dimethyl-1-phenylethylamine 98%
(S)-(-)-N,N-Dimethyl-1-phenylethylamine

Precaución: for Research Use Only

Formula: C₁₀H₁₅N
MW: 149.24 g/mol
MDL Number: MFCD00056236
CAS Number: 17279-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR