3-Nitrosalicylic acid 98%

Supplier: Ambeed
Warning

A430911-100G A430911-1G A430911-25G A430911-5G
77143-272EA 202.91 USD
77143-272 77143-274 77143-276 77143-278
3-Nitrosalicylic acid 98%
3-Nitrosalicylic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₅
MDL Number: MFCD00024240
CAS Number: 85-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR