trans-3-Chlorocinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: (E)-3-(3-Chlorophenyl)acrylic acid

A430275-100G A430275-25G
77143-266EA 124.13 USD
77143-266 77143-268
trans-3-Chlorocinnamic acid 98%
trans-3-Chlorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₇ClO₂
MW: 182.6037 g/mol
MDL Number: MFCD00004384
CAS Number: 14473-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR