5-Fluoro-2,3-dimethylbenzaldehyde 97%

Supplier: Ambeed
Warning

A429591-1G A429591-250MG
77143-262EA 393.6 USD
77143-262 77143-264
5-Fluoro-2,3-dimethylbenzaldehyde 97%
5-Fluoro-2,3-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO
MW: 152.16 g/mol
MDL Number: MFCD28404958
CAS Number: 1803816-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR