[1,1':4',1''-Terphenyl]-4-ylboronic acid 97%
Supplier: Ambeed
|
Warning
|
A429240-1G
A429240-250MG
A429240-5G
77143-256EA
22.65
USD
77143-256
77143-258
77143-260
[1,1':4',1''-Terphenyl]-4-ylboronic acid 97%
[1,1':4',1''-Terphenyl]-4-ylboronic acid
Precaución: for Research Use Only
Formula:
C₁₈H₁₅BO₂ |
MDL Number:
MFCD28133449 CAS Number: 877993-09-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...