2-Chloro-7-cyclopentyl-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylic acid 97%

Supplier: Ambeed
Warning

A428777-100MG A428777-1G A428777-250MG A428777-5G
77143-248EA 26.53 USD
77143-248 77143-250 77143-252 77143-254
2-Chloro-7-cyclopentyl-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylic acid 97%
2-Chloro-7-cyclopentyl-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClN₃O₂
MW: 265.70 g/mol
MDL Number: MFCD29041989
CAS Number: 1211443-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR