N-Ethyl-N-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 97%

Supplier: Ambeed
Warning

A428632-100MG A428632-1G A428632-250MG
77143-236EA 38.12 USD
77143-236 77143-238 77143-240
N-Ethyl-N-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 97%
N-Ethyl-N-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₆H₂₄BNO₃
MW: 289.18 g/mol
CAS Number: 2304635-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR