AVN-944 99%

Supplier: Ambeed
Warning

Synonyms: VX-944, (R)-1-cyanobutan-2-yl ((S)-1-(3-(3-(3-methoxy-4-(oxazol-5-yl)phenyl)ureido)phenyl)ethyl)carbamate

A428299-100MG A428299-250MG A428299-50MG
77143-230EA 509.24 USD
77143-230 77143-232 77143-234
AVN-944 99%
AVN-944

Precaución: for Research Use Only

Formula: C₂₅H₂₇N₅O₅
MW: 477.51 g/mol
MDL Number: MFCD22200574
CAS Number: 297730-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR