2-Bromo-4-hydroxynicotinaldehyde 97%

Supplier: Ambeed
Warning

A426873-100MG A426873-1G A426873-250MG
77143-220EA 52.53 USD
77143-220 77143-222 77143-224
2-Bromo-4-hydroxynicotinaldehyde 97%
2-Bromo-4-hydroxynicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₄BrNO₂
MDL Number: MFCD22547753
CAS Number: 1289132-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR