(2S,3R)-2-(3,4-Dihydroxyphenyl)chroman-3,5,7-triol 98%

Supplier: Ambeed
Warning

Synonyms: (-)-Catechin

A426574-1G A426574-25G
77143-216EA 47.39 USD
77143-216 77143-218
(2S,3R)-2-(3,4-Dihydroxyphenyl)chroman-3,5,7-triol 98%
(2S,3R)-2-(3,4-Dihydroxyphenyl)chroman-3,5,7-triol

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₆
MDL Number: MFCD00135997
CAS Number: 18829-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR