4-[(Boc-amino)methyl]tetrahydro-2H-pyran-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(((tert-Butoxycarbonyl)amino)methyl)tetrahydro-2H-pyran-4-carboxylic acid

A424163-100MG A424163-1G A424163-250MG A424163-5G
77143-206EA 123.6 USD
77143-206 77143-208 77143-210 77143-212
4-[(Boc-amino)methyl]tetrahydro-2H-pyran-4-carboxylic acid 97%
4-[(Boc-amino)methyl]tetrahydro-2H-pyran-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₅
MW: 259.299 g/mol
MDL Number: MFCD09264199
CAS Number: 946761-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR