2-Bromo-5-chloro-3-fluorophenol 95%

Supplier: Ambeed
Warning

A423620-1G A423620-250MG
77143-202EA 160.2 USD
77143-202 77143-204
2-Bromo-5-chloro-3-fluorophenol 95%
2-Bromo-5-chloro-3-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrClFO
MW: 225.44 g/mol
MDL Number: MFCD28734032
CAS Number: 1805479-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR