2-(trans-4-Aminocyclohexyl)acetonitrile 97%

Supplier: Ambeed
Warning

A423578-250MG A423578-1G A423578-100MG A423578-5G
77143-200EA 180.36 USD
77143-200 77551-278 77686-165 77686-166
2-(trans-4-Aminocyclohexyl)acetonitrile 97%
2-(trans-4-Aminocyclohexyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₁₄N₂
MW: 138.21 g/mol
MDL Number: MFCD27942777
CAS Number: 1266328-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR