7-Chloro-5-methoxy-1H-pyrrolo[2,3-c]pyridine 97%
Supplier: Ambeed
|
Warning
|
A423562-100MG
A423562-250MG
77143-196EA
159.14
USD
77143-196
77143-198
7-Chloro-5-methoxy-1H-pyrrolo[2,3-c]pyridine 97%
7-Chloro-5-methoxy-1H-pyrrolo[2,3-c]pyridine
Precaución: for Research Use Only
Formula:
C₈H₇ClN₂O MW: 182.61 g/mol |
MDL Number:
MFCD11845543 CAS Number: 930790-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...