Flumazenil 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 8-fluoro-5-methyl-6-oxo-5,6-dihydro-4H-benzo[f]imidazo[1,5-a][1,4]diazepine-3-carboxylate

A423241-100MG A423241-10MG A423241-1MG A423241-50MG A423241-5MG A423241-1G
77143-180EA 57.29 USD
77143-180 77143-182 77143-184 77143-186 77143-188 77686-164
Flumazenil 95%
Flumazenil

Precaución: for Research Use Only

Formula: C₁₅H₁₄FN₃O₃
MW: 303.29 g/mol
MDL Number: MFCD00242764
CAS Number: 78755-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR